missing translation for 'onlineSavingsMsg'
Learn More

Triethylene Glycol Dimethacrylate, SPEX CertiPrep™

Product Code. 12929404 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12929404 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12929404 Supplier SPEX Certiprep Supplier No. S3704

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Innehåller

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Triethylene glycol dimethacrylate109-16-01000μg/mL

Certifikat

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Uppfyllande

GHS/OSHA Complaint

Specifikationer

Packaging 2mL amber prescored glass ampule
CAS 109-16-0, 67-56-1
CAS Min % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molecular Formula C14H22O6
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key HWSSEYVMGDIFMH-UHFFFAOYSA-N
Type Certified Reference Material
SMILES CC(=C)C(=O)OCCOCCOCCOC(=O)C(=C)C
IUPAC Name 2-[2-[2-(2-methylprop-2-enoyloxy)ethoxy]ethoxy]ethyl 2-methylprop-2-enoate
Solvent or Matrix Methanol-P and T
Molecular Weight (g/mol) 286.324
PubChem CID 7979
Quantity 1 mL
Chemical Name or Material Triethylene glycol dimethacrylate
Visa mer Visa mindre
compliance-icons compliance-icons compliance-icons
Produktbeteckning
  • Triethylene glycol dimethacrylate
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Faroangivelser
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Skyddsangivelse
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Kompletterande information
  • MIXTURE LIST-Contain: methanol

Product intended for laboratory use only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.