missing translation for 'onlineSavingsMsg'
Learn More

Triethylene glycol dimethacrylate, contains MEHQ as inhibitor

Product Code. 17231156 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17231156 1 L 1 L
17241156 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17231156 Supplier Thermo Scientific Acros Supplier No. 466270010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-16-0 | C14H22O6 | 286.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-16-0
Molecular Formula C14H22O6
Molecular Weight (g/mol) 286.32
InChI Key HWSSEYVMGDIFMH-UHFFFAOYSA-N
IUPAC Name 2-(2-{2-[(2-methylprop-2-enoyl)oxy]ethoxy}ethoxy)ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCOCCOCCOC(=O)C(C)=C

Specifications

Density 1.0920 g/mL
Boiling Point 170.0°C to 172.0°C (5.0 mmHg)
Flash Point 167°C
Quantity 1 L
Specific Gravity 1.092
Solubility Information Solubility aq. soln.: 3.6 g/L (20°C). Other solubilities: miscible with most organic solvents
Viscosity 11 mPa.s (25°C)
Formula Weight 286.32
Chemical Name or Material Triethylene glycol dimethacrylate
compliance-icons
Product Identifier
  • Triethylene glycol dimethacrylate, contains MEHQ as inhibitor
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Triethylene glycol dimethacrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.