missing translation for 'onlineSavingsMsg'
Learn More

Triethylene glycol diacetate, 98%, Thermo Scientific™

Product Code. p-7046012
Click to view available options
Quantity:
250 g
Unit Size:
250g
This item is not returnable. View return policy

Product Code. 11393366

Brand: Thermo Scientific Alfa Aesar L12789.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-21-7
Molecular Formula C10H18O6
Molecular Weight (g/mol) 234.248
MDL Number MFCD00026213
InChI Key OVOUKWFJRHALDD-UHFFFAOYSA-N
Synonym triethylene glycol diacetate, tdac, triglycol diacetate, triethylene glycol, diacetate, triglycol, diacetate, acetic acid, triethylene glycol diester, triethyleneglycoldiacetate, 2,2'-ethylenedioxy di ethyl acetate, ethanol, 2,2'-ethylenedioxydi-, diacetate, ethanol, 2,2'-1,2-ethanediylbis oxy bis-, diacetate
PubChem CID 8098
IUPAC Name 2-[2-(2-acetyloxyethoxy)ethoxy]ethyl acetate
SMILES CC(=O)OCCOCCOCCOC(=O)C

Specifications

Melting Point -50°C
Density 1.12
Boiling Point 286°C
Flash Point 163°C (325°F)
Refractive Index 1.44
Quantity 250 g
Beilstein 1789453
Formula Weight 234.25
Percent Purity 98%
Chemical Name or Material Triethylene glycol diacetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.