missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 2-phosphonobutyrate, 98%, Thermo Scientific Chemicals

Product Code. 10243052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10243052 25 mL Glass bottle 25mL
10702843 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10243052 Supplier Thermo Scientific Acros Supplier No. 368780250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17145-91-4
Molecular Formula C10H21O5P
Molecular Weight (g/mol) 252.25
MDL Number MFCD00041347
InChI Key GYUCVQSNZFRDRL-UHFFFAOYSA-N
Synonym triethyl 2-phosphonobutyrate, ethyl 2-diethoxyphosphoryl butanoate, ethyl 2-diethylphosphono butyrate, butanoic acid, 2-diethoxyphosphinyl-, ethyl ester, triethyl 2-phosphonobutanoate, .alpha.-diethylphosphono butanoic acid, ethyl ester, butyric acid, 2-phosphono-, triethyl ester, butyric acid, triethyl ester, triethyl 2-phosphonobu-tyrate, triethyl-2-phosphono butyrate
PubChem CID 229053
IUPAC Name ethyl 2-diethoxyphosphorylbutanoate
SMILES CCC(C(=O)OCC)P(=O)(OCC)OCC

Specifications

Color Colorless
Density 1.0640g/mL
Boiling Point 152.0°C to 154.0°C (14.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (O)P(CH3CH2O)2CH(CH2CH3)C(O)OCH2CH3
Refractive Index 1.4315 to 1.4335
Quantity 25 mL
Specific Gravity 1.064
Formula Weight 252.25
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Triethyl 2-phosphonobutyrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Triethyl 2-phosphonobutyrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.