missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Triethylammonium Formate Solution, Honeywell™

Product Code. 15611600 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15611600 500 mL 500mL
15601600 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15611600 Supplier Honeywell Fluka™ Supplier No. 17901500ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

volatile buffer, 1 M pH 6.0, for HPLC

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 585-29-5
Color Colorless
For Use With (Equipment) Volatile buffer for HPLC
pH 5.9 to 6.1 (at 25°C)
Physical Form Liquid
Quantity 500 mL
Packaging Plastic Bottle
Molecular Weight (g/mol) 147.218
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.