missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Triethylammonium Bicarbonate Buffer, Honeywell™ Fluka™

Product Code. 15658320 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 15658320

Brand: Honeywell Fluka™ 17902500ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

volatile buffer, for HPLC, 1 M

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 15715-58-9
Color Colorless
For Use With (Equipment) Volatile buffer for HPLC
pH 8.4 to 8.6 (at 25°C)
Physical Form Liquid
Quantity 500 mL
Packaging Glass Bottle
Molecular Weight (g/mol) 163.217
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.