missing translation for 'onlineSavingsMsg'
Learn More

Solstice Triethylammonium acetate buffer, BioUltra, volatile buffer, ∼1.0 M in H2O, Solstice

Product Code. 15647420
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15647420 500 mL 500mL
15637420 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15647420 Supplier Solstice Supplier No. 90357500ML
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 24-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BioUltra, volatile buffer, ∼1.0 M in H2O

  • Finest quality products for life science
  • The most stringent testing protocols and highest purity
  • Superior manufacturing methods and product specific testing protocols designed to ensure quality to our highest standard
  • Tests may include, but are not limited to, purity by gel electrophoresis, trace metal testing and application testing.
TRUSTED_SUSTAINABILITY

Specifications

CAS 5204-74-0
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.