missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, Certified AR for Analysis, Fisher Chemical™

Product Code. 10704284 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
Unit Size:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10704284 25 L Metal can 25 L
10224020 1 L Amber glass bottle 1 L
10417152 2.5 L Plastic Coated Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Product Code. 10704284 Supplier Fisher Chemical Supplier No. T/3201/25
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Triethylamine, Certified AR for Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
MDL Number 9051
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC

Specifications

Melting Point -115°C
Color Colorless
pH 12.4
Boiling Point 90°C
Packaging Metal can
Quantity 25 L
Vapor Density 3.5
Viscosity 0.36 mPaS at 20°C
Formula Weight 101.19g/mol
Vapor Pressure 69mbar at 20°C
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.