missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog Number p-5383293
270.75 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
121-44-8
C6H15N
101.193
ZMANZCXQSJIPKH-UHFFFAOYSA-N
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
8471
CHEBI:35026
N,N-diethylethanamine
CCN(CC)CC
This item is not returnable. View return policy
121-44-8
C6H15N
101.193
ZMANZCXQSJIPKH-UHFFFAOYSA-N
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
8471
CHEBI:35026
N,N-diethylethanamine
CCN(CC)CC

High quality chemicals for laboratory and specialized industrial use

Flash Point −11.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 500 mL
Specific Gravity 1L = 0.73kg
Formula Weight 101.19
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Triethylamine
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'