missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, ≥99.5%, Solstice

Product Code. 15638500 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 x 2 mL
Packaging:
Ampoule
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15638500 10 x 2 mL Ampoule Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15638500 Supplier Honeywell Fluka Supplier No. 1792410X2ML
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 121-44-8
Density 0.726g/mL at 25°C
Explosion Limit 0.08
Flash Point −15°C (5°F)
Ignition Point 593°F
Packaging Ampoule
Molecular Formula C6H15N
Linear Formula (C2H5)3N
MDL Number MFCD00009051
Quantity 10 x 2 mL
UN Number UN1296
Beilstein 605283
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
SMILES CCN(CC)CC
IUPAC Name N,N-diethylethanamine
Molecular Weight (g/mol) 101.193
PubChem CID 8471
Vapor Density 3.5 (vs air)
ChEBI CHEBI:35026
Formula Weight 101.19g/mol
Vapor Pressure 51.75mmHg (20°C)
Percent Purity ≥99.5%
Chemical Name or Material Triethylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.