missing translation for 'onlineSavingsMsg'
Learn More

Triethylaluminum, 94%, Thermo Scientific Chemicals

Product Code. 15431877
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15431877 25 g 25g
15441877 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15431877 Supplier Thermo Scientific Alfa Aesar Supplier No. 043243.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Catalyst for polymerization of olefins, intermediate

Triethylaluminum is used as a co-catalyst in the industrial production of polyethylene and for the production of medium chain alcohols. Used as a catalyst in Ziegler-Natta polymerization process for vinyl, olefin, diene polymerizations and linear oligomerization and cyclization of unsaturated hydrocarbons. It is also used as a catalyst to produce ethylene gas, chain growth of ethylene, longer chain aluminum alkyls, and in plating aluminum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-93-8
Molecular Formula C6H15Al
Molecular Weight (g/mol) 114.168
MDL Number MFCD00009015
InChI Key VOITXYVAKOUIBA-UHFFFAOYSA-N
Synonym triethylaluminum, triethylaluminium, aluminum, triethyl, triethylalane, unii-h426e9h3tt, triethylaluminum solution, 1.0 m in hexanes, triethylaluminum solution, 25 wt. % in toluene, triethyl-aluminum, triethyl aluminum, triethyl-aluminum
PubChem CID 16682930
IUPAC Name triethylalumane
SMILES CC[Al](CC)CC

Specifications

Melting Point -50°C
Density 0.835g/mL
Boiling Point 128°C to 130°C (50 mmHg)
Flash Point 18°C (−1°F)
Odor Characteristic
Packaging Cylinder
Quantity 25 g
UN Number UN3394
Sensitivity Moisture Sensitive
Formula Weight 114.17
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Triethylaluminum
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.