missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyl Orthoformate, TRC
Click to view available options
Quantity:
500 g
Unit Size:
500g
Specifications
Specifications
| Chemical Name or Material | Triethyl Orthoformate |
| CAS | 122-51-0 |
| Molecular Formula | C7H16O3 |
| Quantity | 500 g |
| Synonym | 1,1',1''-[Methylidyne-tris(oxy)]tris-Ethane, Orthoformic Acid Triethyl Ester |
| InChI Formula | InChI=1S/C7H16O3/c1-4-8-7(9-5-2)10-6-3/h7H,4-6H2,1-3H3 |
| SMILES | CCOC(OCC)OCC |
| IUPAC Name | diethoxymethoxyethane |
| Molecular Weight (g/mol) | 148.2 |
| Formula Weight | 148.1099 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction