missing translation for 'onlineSavingsMsg'
Learn More

Triethyl orthoformate, Reagent Grade, 98%, Honeywell™

Product Code. 15688180 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
20 L
Packaging:
Glass Bottle
Stainless steel can
Unit Size:
1 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15688180 1 L Glass Bottle 1 L
15698180 20 L Stainless steel can 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15688180 Supplier Honeywell Research Chemicals Supplier No. T604531L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 122-51-0
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.202
InChI Key GKASDNZWUGIAMG-UHFFFAOYSA-N
Synonym triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty
PubChem CID 31214
IUPAC Name diethoxymethoxyethane
SMILES CCOC(OCC)OCC

Specifications

Melting Point -76°C (literature)
Density 0.891 g/mL at 25 °C(lit.)
Boiling Point 146°C (literature)
Packaging Glass Bottle
Linear Formula CH(OC2H5)3
MDL Number MFCD00131528
Quantity 1 L
UN Number UN2524
Beilstein 605384
Vapor Density 5.11 (vs air)
Percent Purity >97.5%
Grade Reagent
Chemical Name or Material Triethyl orthoformate, Reagent grade, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.