missing translation for 'onlineSavingsMsg'
Learn More

Triethyl orthoformate, 98%

Product Code. 11400124
Change view
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
Unit Size:
10000mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400124 2500 mL 2500mL
11490114 500 mL 500mL
11410124 10,000 mL 10000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11400124 Supplier Thermo Scientific Alfa Aesar Supplier No. A13587.0F
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 122-51-0 | C7H16O3 | 148.202 g/mol

Triethyl orthoformate is used in Bodroux-Chichibabin aldehyde synthesis to prepare an aldehyde with one carbon higher by reacting with Grignard reagent. It is also used in the electrophilic formylation of activated aromatic species such as phenol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl orthoformate is used in Bodroux-Chichibabin aldehyde synthesis to prepare an aldehyde with one carbon higher by reacting with Grignard reagent. It is also used in the electrophilic formylation of activated aromatic species such as phenol.

Solubility
Miscible with alcohols, acetone, ethers and chloroform. Slightly miscible with water.

Notes
Moisture sensitive. Incompatible with acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-51-0
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.202
MDL Number MFCD00009230
InChI Key GKASDNZWUGIAMG-UHFFFAOYSA-N
Synonym triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty
PubChem CID 31214
IUPAC Name diethoxymethoxyethane
SMILES CCOC(OCC)OCC

Specifications

Melting Point -61°C
Density 0.891
Boiling Point 144°C to 146°C
Flash Point 30°C (86°F)
Assay Percent Range 98%
Linear Formula HC(OCH2CH3)3
Refractive Index 1.391
Quantity 2500 mL
UN Number UN2524
Beilstein 605384
Sensitivity Moisture sensitive
Merck Index 14,6884
Solubility Information Miscible with alcohols,acetone,ethers and chloroform. Slightly miscible with water.
Formula Weight 148.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Triethyl orthoformate
Show More Show Less
compliance-icons
Product Identifier
  • Triethyl orthoformate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: Triethyl orthoformate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.