missing translation for 'onlineSavingsMsg'
Learn More

Triethyl orthoformate, 98%

Product Code. 10297133 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
Unit Size:
1 L
100mL
2.5 L
25 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10297133 25 L Metal Drum 25 L
10512541 100 mL Glass bottle 100mL
10476960 1 L Glass bottle 1 L
10225430 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10297133 Supplier Thermo Scientific Acros Supplier No. 139660250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 122-51-0 | C7H16O3 | 148.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-51-0
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.2
MDL Number MFCD00009230
InChI Key GKASDNZWUGIAMG-UHFFFAOYSA-N
Synonym triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty
PubChem CID 31214
IUPAC Name diethoxymethoxyethane
SMILES CCOC(OCC)OCC

Specifications

Melting Point -61.0°C
Color Colorless
Density 0.8900g/mL
Boiling Point 146.0°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Metal Drum
Linear Formula CH(OC2H5)3
Refractive Index 1.3900 to 1.3920
Quantity 25 L
Beilstein 02, 20
Fieser 01,1204; 02,430; 04,527; 05,690; 06,610; 07,385; 11,555; 13,327
Merck Index 15, 6980
Specific Gravity 0.89
Solubility Information Solubility in water: 1.35g/L (20°C). Other solubilities: decomposes in water,soluble in alcohols,acetone,ethers,chloroform,
Formula Weight 148.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Triethyl orthoformate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.