missing translation for 'onlineSavingsMsg'
Learn More

Triethyl orthoformate, 98%, Thermo Scientific Chemicals

Product Code. 10476960
Click to view available options
Quantity:
100 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-51-0
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.2
MDL Number MFCD00009230
InChI Key GKASDNZWUGIAMG-UHFFFAOYSA-N
Synonym triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty
PubChem CID 31214
IUPAC Name diethoxymethoxyethane
SMILES CCOC(OCC)OCC

Specifications

Melting Point -61.0°C
Color Colorless
Density 0.8900g/mL
Boiling Point 146.0°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH(OC2H5)3
Refractive Index 1.3900 to 1.3920
Quantity 1 L
Beilstein 02, 20
Fieser 01,1204; 02,430; 04,527; 05,690; 06,610; 07,385; 11,555; 13,327
Merck Index 15, 6980
Specific Gravity 0.89
Solubility Information Solubility in water: 1.35g/L (20°C). Other solubilities: decomposes in water,soluble in alcohols,acetone,ethers,chloroform,
Formula Weight 148.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Triethyl orthoformate
Show More Show Less
compliance-icons
Product Identifier
  • Triethyl orthoformate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'