missing translation for 'onlineSavingsMsg'
Learn More

Triethyl orthobenzoate, 97%

Product Code. 10401553 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10401553 25 g Glass bottle 25g
10177832 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10401553 Supplier Thermo Scientific Acros Supplier No. 378760250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1663-61-2 | C13H20O3 | 224.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1663-61-2
Molecular Formula C13H20O3
Molecular Weight (g/mol) 224.3
MDL Number MFCD00009222
InChI Key BQFPCTXLBRVFJL-UHFFFAOYSA-N
Synonym triethoxymethyl benzene, triethyl orthobenzoate, ethyl orthobenzoate, benzene, triethoxymethyl, triethylorthobenzoate, orthobenzoic acid, triethyl ester, orthobenzoic acid triethyl ester, triethoxymethyl benzol, triethoxyphenylmethane
PubChem CID 74268
IUPAC Name triethoxymethylbenzene
SMILES CCOC(C1=CC=CC=C1)(OCC)OCC

Specifications

Color Colorless
Density 0.9910g/mL
Boiling Point 239°C to 241°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C(OCH2CH3)3
Refractive Index 1.4710 to 1.474
Quantity 25 g
Specific Gravity 0.991
Solubility Information Solubility in water: hydrolysis
Formula Weight 224.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Triethyl orthobenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Triethyl orthobenzoate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.