missing translation for 'onlineSavingsMsg'
Learn More

Triethyl orthoacetate, 97%

Product Code. 10265380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10265380 100 mL 100mL
10061510 500 mL 500mL
10185683 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10265380 Supplier Thermo Scientific Acros Supplier No. 139651000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-39-7 | C8H18O3 | 162.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-39-7
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.23
MDL Number MFCD00009223
InChI Key NDQXKKFRNOPRDW-UHFFFAOYSA-N
Synonym triethyl orthoacetate, ethyl orthoacetate, ethane, 1,1,1-triethoxy, orthoacetic acid, triethyl ester, orthoacetic acid triethyl ester, triethoxyethane, triethoxy ethane, triethy orthoacetate, triethyl ortho-aceate, 1,1-triethoxyethane
PubChem CID 66221
IUPAC Name 1,1,1-triethoxyethane
SMILES CCOC(C)(OCC)OCC

Specifications

Color Colorless to Yellow
Density 0.8850g/mL
Boiling Point 142.0°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3C(OC2H5)3
Refractive Index 1.3950 to 1.3980
Quantity 100 mL
Beilstein 02, 129
Fieser 03,300; 04,527; 06,607; 10,417
Specific Gravity 0.885
Solubility Information Solubility in water: decomposes
Viscosity 1 mPa.s (20°C)
Formula Weight 162.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Triethyl orthoacetate, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triethyl orthoacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Triethyl orthoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.