missing translation for 'onlineSavingsMsg'
Learn More

Triethyl methanetricarboxylate, 98%

Product Code. 11368105
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11368105

Brand: Thermo Scientific Alfa Aesar L08356.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6279-86-3 | C10H16O6 | 232.232 g/mol

Methanetricarboxylates have been used as blocked malonic esters, restricting reaction with alkyl halides to monoalkylation. The unwanted carboxyl group can be removed by treatment with Na alkoxide, LDA or BCl3. Provides a useful 2-carbon chain extension method that adds to Michael acceptors under phase-transfer conditions. Triethyl Methanetricarboxylate is used in the synthesis of novel inhibitors of Hsp90. It is also used in the preparation of novel dihydroquinoline-3-carboxylic acids that function as HIV-1 integrase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methanetricarboxylates have been used as ″blocked″ malonic esters, restricting reaction with alkyl halides to monoalkylation. The unwanted carboxyl group can be removed by treatment with Na alkoxide, LDA or BCl3. Provides a useful 2-carbon chain extension method that adds to Michael acceptors under phase-transfer conditions. Triethyl Methanetricarboxylate is used in the synthesis of novel inhibitors of Hsp90. It is also used in the preparation of novel dihydroquinoline-3-carboxylic acids that function as HIV-1 integrase inhibitors.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6279-86-3
Molecular Formula C10H16O6
Molecular Weight (g/mol) 232.232
MDL Number MFCD00009150
InChI Key AGZPNUZBDCYTBB-UHFFFAOYSA-N
Synonym tricarbethoxymethane, triethylmethanetricarboxylate, methanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-malonic acid diethyl ester, methanetricarboxylic acid triethyl ester, carboxymalonic acid triethyl ester, diethyl 2-ethoxycarbonyl malonate, diethyl 2-ethoxycarbonyl propane-1,3-dioate, pubchem3106, triethoxycarbonylmethane
PubChem CID 80471
IUPAC Name triethyl methanetricarboxylate
SMILES CCOC(=O)C(C(=O)OCC)C(=O)OCC

Specifications

Melting Point 26°C to 29°C
Density 1.095
Boiling Point 253°C
Flash Point >110°C (230°F)
Refractive Index 1.4255
Quantity 5 g
Beilstein 1793645
Solubility Information Insoluble in water.
Formula Weight 232.23
Percent Purity 98%
Chemical Name or Material Triethyl methanetricarboxylate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.