missing translation for 'onlineSavingsMsg'
Learn More

Triethyl methanetricarboxylate, 98%

Product Code. 10793891 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 10793891

Brand: Thermo Scientific Acros 139630250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6279-86-3 | C10H16O6 | 232.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6279-86-3
Molecular Formula C10H16O6
Molecular Weight (g/mol) 232.23
InChI Key AGZPNUZBDCYTBB-UHFFFAOYSA-N
Synonym tricarbethoxymethane, triethylmethanetricarboxylate, methanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-malonic acid diethyl ester, methanetricarboxylic acid triethyl ester, carboxymalonic acid triethyl ester, diethyl 2-ethoxycarbonyl malonate, diethyl 2-ethoxycarbonyl propane-1,3-dioate, pubchem3106, triethoxycarbonylmethane
PubChem CID 80471
IUPAC Name triethyl methanetricarboxylate
SMILES CCOC(=O)C(C(=O)OCC)C(=O)OCC

Specifications

Melting Point 29.0°C
Color Colorless to Yellow
Density 1.1000g/mL
Boiling Point 253.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH(CO2C2H5)3
Refractive Index 1.4240 to 1.4260
Quantity 25 mL
Beilstein 02,81
Specific Gravity 1.1
Solubility Information Solubility in water: immiscible
Formula Weight 232.23
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Triethyl methanetricarboxylate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.