missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyl 4-phosphonocrotonate, cis + trans, 94%, Thermo Scientific Chemicals
46.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
25 g
250 g
Description
Triethyl 4-phosphonocrotonate is used as a reactant for iminodipropionic acid as a leaving group for DNA polymerization by HIV-1 reverse transcriptase, orally bioavailable GPR40 agonist synthesis, preparation of mGlu4R agonists and synthesis of fluoroketone inhibitors of group VIA calcium-independent phospholipase A2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Triethyl 4-phosphonocrotonate is used as a reactant for iminodipropionic acid as a leaving group for DNA polymerization by HIV-1 reverse transcriptase, orally bioavailable GPR40 agonist synthesis, preparation of mGlu4R agonists and synthesis of fluoroketone inhibitors of group VIA calcium-independent phospholipase A2.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Triethyl 4-phosphonocrotonate is used as a reactant for iminodipropionic acid as a leaving group for DNA polymerization by HIV-1 reverse transcriptase, orally bioavailable GPR40 agonist synthesis, preparation of mGlu4R agonists and synthesis of fluoroketone inhibitors of group VIA calcium-independent phospholipase A2.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Density | 1.13 |
Boiling Point | 133°C to 135°C (0.4 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.455 |
Quantity | 10 g |
Formula Weight | 250.23 |
Percent Purity | 94% |
Chemical Name or Material | Triethyl 4-phosphonocrotonate, cis + trans |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'