missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 2-Phosphonopropionate, TRC

Product Code. 30451194
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451194 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451194 Supplier Toronto Research Chemicals Supplier No. TRCT79811050G
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triethyl 2-Phosphonopropionate
CAS 3699-66-9
Molecular Formula C9H19O5P
Quantity 50 g
Synonym 2-(Diethoxyphosphinyl)propanoic Acid Ethyl Ester, 2-Phosphonopropionic Acid','','Triethyl Ester, 2-(Diethoxyphosphinyl)propanoic Acid Ethyl Ester, 2-(Diethoxyphosphoryl)propionic Acid Ethyl Ester, 2-Phosphonopropanoic Acid Triethyl ester, 2-Phosphonopropionic Acid Triethyl Ester, Ethyl 2-(Diethoxyphosphinyl)propanoate, Ethyl 2-(Diethoxyphosphoryl)propanoate, Ethyl 2-(diethylphosphono)propionate, Ethyl alpha-diethylphosphonopropionate, Triethyl 2-Phosphonopropanoate, Triethyl 2-phosphonopropionate, Triethyl alpha-Phosphonopropionate, alpha-Diethylphosphonopropionic Acid Ethyl Ester
InChI Formula InChI=1S/C9H19O5P/c1-5-12-9(10)8(4)15(11,13-6-2)14-7-3/h8H,5-7H2,1-4H3
SMILES CCOC(=O)C(C)P(=O)(OCC)OCC
IUPAC Name ethyl 2-diethoxyphosphorylpropanoate
Molecular Weight (g/mol) 238.22
Formula Weight 238.097
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.