missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 1,1,2-ethanetricarboxylate, 99%

Product Code. 10627302 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10627302 100 g Glass bottle 100g
10636714 5 g Glass bottle 5g
10285330 25 g Glass bottle 25g
10217213 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10627302 Supplier Thermo Scientific Acros Supplier No. 139611000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7459-46-3 | C11H18O6 | 246.259 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7459-46-3
MDL Number MFCD00009154
InChI Key TVWZLLYAJDSSCJ-UHFFFAOYSA-N
Synonym triethyl 1,1,2-ethanetricarboxylate, 1,1,2-ethanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-succinic acid diethyl ester, triethyl ethane tricarboxylate, 1,1,2-triethyl ethane-1,1,2-tricarboxylate, triethyl ethane-1,2,2-tricarboxylate, ethane-1,1,2-tricarboxylic acid triethyl ester, ethane-1,1,2-tricarboxylic acid, triethyl ester, diethyl 2-ethoxycarbonyl butane-1,4-dioate, acmc-209ovd
PubChem CID 81961
IUPAC Name triethyl ethane-1,1,2-tricarboxylate
SMILES CCOC(=O)CC(C(=O)OCC)C(=O)OCC

Specifikationer

Color Colorless
Density 1.07g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C11H18O6
Linear Formula C2H5O2CCH2CH(CO2C2H5)2
Refractive Index 1.4280 to 1.4300 (20°C, 589nm)
Quantity 100 g
Beilstein 02, 813
Molecular Weight (g/mol) 246.259
Formula Weight 246.26
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Triethyle1, 1, 2-ethanetricarboxylate
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.