missing translation for 'onlineSavingsMsg'
Learn More

Triethoxyvinylsilane, 97%, Thermo Scientific Chemicals

Product Code. p-203828 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
500 g
Packaging:
Glass bottle
Unit Size:
25g
500g
This item is not returnable. View return policy

Product Code. 10498320

Brand: Thermo Scientific Acros 174610250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-08-0
Molecular Formula C8H18O3Si
Molecular Weight (g/mol) 190.31
MDL Number MFCD00009063
InChI Key FWDBOZPQNFPOLF-UHFFFAOYSA-N
Synonym vinyltriethoxysilane, triethoxyvinylsilane, silane, ethenyltriethoxy, triethoxy vinyl silane, polyscience vtes, triethoxyvinylsilicane, vtes, silane a 151, ethenyl triethoxy silane, silane, triethoxyvinyl
PubChem CID 6516
SMILES CCO[Si](OCC)(OCC)C=C

Specifications

Color Colorless
Density 0.9000g/mL
Boiling Point 160°C to 161°C
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHSi(OC2H5)3
Refractive Index 1.3970 to 1.399
Quantity 25 g
Specific Gravity 0.9
Solubility Information Solubility in water: decomposes in water. Other solubilities: soluble in chloroform
IUPAC Name ethenyltriethoxysilane
Formula Weight 190.32
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Triethoxyvinylsilane, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Triethoxyvinylsilane
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : Triethoxyvinylsilane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.