missing translation for 'onlineSavingsMsg'
Learn More

Triethoxysilane, 95%

Product Code. 13428398 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13428398 250 g Glass bottle 250g
10163294 10 g Glass bottle 10g
10366834 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13428398 Supplier Thermo Scientific Acros Supplier No. 164812500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 998-30-1 | C6H16O3Si | 164.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 998-30-1
Molecular Formula C6H16O3Si
Molecular Weight (g/mol) 164.28
MDL Number MFCD00009061
InChI Key PKDCQJMRWCHQOH-UHFFFAOYSA-N
Synonym triethoxysilane, silane, triethoxy, unii-8t460wdh89, c6h16o3si, si oet 3h;, c2h5o 3sih, 4-01-00-01359 beilstein handbook reference, triethoxysilane 10g, triethoxysilyl modified poly 1,2-butadiene, poly 1,2-butadiene , triethoxysilyl modified
PubChem CID 6327710
IUPAC Name triethoxysilicon
SMILES CCO[Si](OCC)OCC

Specifications

Melting Point -170.0°C
Color Colorless to Yellow
Density 0.8900g/mL
Boiling Point 134.0°C to 135.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5O)3SiH
Refractive Index 1.3762
Quantity 250 g
Beilstein 01,334
Fieser 11,554; 17,366
Specific Gravity 0.89
Solubility Information Solubility in water: reacts
Formula Weight 164.28
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Triethoxysilane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Triethoxysilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.