missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tridecanoic-d25 Acid, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Tridecanoic-d25 Acid |
| CAS | 202529-03-1 |
| Molecular Formula | C13 D25 H O2 |
| Quantity | 50 mg |
| Synonym | NSC 25955-d25, NSC 69131-d25, Tridecylic-d25 Acid, n-Tridecanoic-d25 Acid, n-Tridecoic-d25 Acid, Tridecanoic-d25 acid (9CI), Tridecanoic acid-d25, Perdeuterotridecylic acid, Tridecanoic-D25 acid |
| InChI Formula | InChI=1S/C13H26O2/c1-2-3-4-5-6-7-8-9-10-11-12-13(14)15/h2-12H2,1H3,(H,14,15)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,13-pentacosadeuteriotridecanoic acid |
| Molecular Weight (g/mol) | 239.5 |
| Formula Weight | 239.3502 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction