missing translation for 'onlineSavingsMsg'
Learn More

Tricyclohexyltin chloride, 97%

Product Code. 15481768
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15481768 25 g 25g
15461768 1 g 1g
15471768 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15481768 Supplier Thermo Scientific Alfa Aesar Supplier No. H55493.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3091-32-5 | C18H33ClSn | 403.622 g/mol

Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.

Solubility
Soluble in ethanol, ether, toluene and chloroform. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3091-32-5
Molecular Formula C18H33ClSn
Molecular Weight (g/mol) 403.622
MDL Number MFCD00003817
InChI Key OJVZYIKTLISAIH-UHFFFAOYSA-M
Synonym tricyclohexyltin chloride, stannane, chlorotricyclohexyl, chlorotricyclohexyltin, chloro tricyclohexyl stannane, tin, chlorotricyclohexyl, tricyclohexyltinchloride, acmc-1ck6z, tricyclohexylchlorotin
PubChem CID 76539
IUPAC Name chloro(tricyclohexyl)stannane
SMILES C1CCC(CC1)[Sn](C2CCCCC2)(C3CCCCC3)Cl

Specifications

Melting Point 123°C to 125°C
Linear Formula (C6H11)3SnCl
Quantity 25 g
UN Number UN3146
Solubility Information Soluble in ethanol,ether,toluene and chloroform. Insoluble in water.
Formula Weight 403.6
Percent Purity 97%
Chemical Name or Material Tricyclohexyltin chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.