missing translation for 'onlineSavingsMsg'
Learn More

Tricyclohexylphosphine, technical grade

Product Code. 11306269
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11306269 1 g 1g
11316269 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11306269 Supplier Thermo Scientific Alfa Aesar Supplier No. 044854.03
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2622-14-2 | C18H33P | 280.436 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2622-14-2
Molecular Formula C18H33P
Molecular Weight (g/mol) 280.436
MDL Number MFCD00003853
InChI Key WLPUWLXVBWGYMZ-UHFFFAOYSA-N
Synonym tricyclohexylphosphine, tricyclohexyl phosphine, phosphine, tricyclohexyl, tricyclohexyl-phosphane, pcy3, tricyclohexylphosphine solution, tchp, p cy 3, phosphorus tricyclohexyl, tricyclohexylphosphine, dissolved in toluene concentration
PubChem CID 75806
IUPAC Name tricyclohexylphosphane
SMILES C1CCC(CC1)P(C2CCCCC2)C3CCCCC3

Specifications

Melting Point 76°C to 78°C
Color White
Odor Unpleasant
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 280.42
Grade Technical
Physical Form Solid
Chemical Name or Material Tricyclohexylphosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.