missing translation for 'onlineSavingsMsg'
Learn More

Tricyclohexylphosphine oxide

Product Code. 11377258
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377258 25 g 25g
11397258 1 g 1g
11387258 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11377258 Supplier Thermo Scientific Alfa Aesar Supplier No. 030387.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13689-19-5 | C18H33OP | 296.44 g/mol

Tricyclohexylphosphine oxide is used as a ligand for a range of metal-catalyzed organic transformations. Ligand used in the Pd-catalyzed coupling of malononitrile with aryl halides.1 Bulky phosphine ligand used with a Pd(0)-triolefinic macrocycle catalyst for Suzuki coupling of aryl bromides and chlorides. This ligand is applied with Ni as a key intermediate in the formation of cylcopentane compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13689-19-5
Molecular Formula C18H33OP
Molecular Weight (g/mol) 296.44
MDL Number MFCD00014298
InChI Key LEFPWWWXFFNJAA-UHFFFAOYSA-N
Synonym tricyclohexylphosphine oxide, phosphine oxide, tricyclohexyl, dicyclohexylphosphoroso cyclohexane, tricyclohexylphosphineoxide, acmc-1bo3x, 4-16-00-01009 beilstein handbook reference, tricyclohexylphosphine oxide 1g
PubChem CID 26187
IUPAC Name dicyclohexylphosphorylcyclohexane
SMILES O=P(C1CCCCC1)(C1CCCCC1)C1CCCCC1

Specifications

Melting Point 155°C to 157°C
Quantity 25 g
UN Number UN3464
Solubility Information Soluble in most organic solvents. Insoluble in water.
Formula Weight 296.42
Physical Form Crystalline
Chemical Name or Material Tricyclohexylphosphine oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.