missing translation for 'onlineSavingsMsg'
Learn More

Tricyclohexylphosphine, 96%, Thermo Scientific Chemicals

Product Code. 11367258
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tricyclohexylphosphine is used as a ligand in organometallic chemistry. As a reagent, it is used for a range of metal-catalyzed organic transformations. Crabtree's catalyst is an organoiridium compound containing tricyclohexyl phosphine used for hydrogenation of mono-, di-, tri-, and tetra- substituted substrates and hydrogen transfer reactions. Grubbs’catalysts consists of transition metal ruthenium and tricylohexyl phosphine utilized in olefin metathesis in synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2622-14-2
Molecular Formula C18H33P
Molecular Weight (g/mol) 280.436
MDL Number MFCD00003853
InChI Key WLPUWLXVBWGYMZ-UHFFFAOYSA-N
Synonym tricyclohexylphosphine, tricyclohexyl phosphine, phosphine, tricyclohexyl, tricyclohexyl-phosphane, pcy3, tricyclohexylphosphine solution, tchp, p cy 3, phosphorus tricyclohexyl, tricyclohexylphosphine, dissolved in toluene concentration
PubChem CID 75806
IUPAC Name tricyclohexylphosphane
SMILES C1CCC(CC1)P(C2CCCCC2)C3CCCCC3

Specifications

Melting Point 76°C to 78°C
Color White
Odor Unpleasant
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in most organic solvents. Insoluble in water.
Formula Weight 280.42
Percent Purity 96%
Physical Form Solid
Chemical Name or Material Tricyclohexylphosphine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'