missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Tricyclic cytosine tC
GREENER_CHOICE

Product Code. 30000442
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000442 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000442 Supplier Tocris Bioscience™ Supplier No. 7814/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorescent tricyclic cytosine analog for RNA imaging

Tricyclic cytosine tC is a fluorescent cytosine analog for imaging RNA in living cells. Enables the investigation of RNA synthesis, degradation, and trafficking at single-cell resolution when combined with confocal imaging. Excitation/emission maxima (λ) = 375/505 nm; extinction coefficient = 4000 M-1 cm-1; quantum yield = 0.2.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-β-D-Ribofuranosyl-1H-pyrimido[5,4-b][1,4]benzothiazin-2(3H)-one
CAS 1174063-74-1
Quantity 10 mg
Target Fluorescent RNA Probes
Molecular Formula C15H15N3O5S
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.