missing translation for 'onlineSavingsMsg'
Learn More

Tricosanol, 99+%

Product Code. 10267490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10267490 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10267490 Supplier Thermo Scientific Acros Supplier No. 258161000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3133-01-5 | C23H48O | 340.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3133-01-5
Melting Point 74°C to 76°C
Color White
Boiling Point 185°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C23H48O
Linear Formula CH3(CH2)22OH
MDL Number MFCD00044484
Quantity 100 mg
Synonym 1-tricosanol, tricosanol, tricosyl alcohol, n-tricosanol, unii-k22080opox, n-tricosyl alcohol, acmc-2097cw, 1-tricosanol 10 microg/ml in methyl-tert. butyl ether
Solubility Information Solubility in water: .. Other solubilities: soluble in hydrocarbons,alcohols,ethers,ketones,specially when hot
InChI Key FPLNRAYTBIFSFW-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCCCCCCCCO
IUPAC Name tricosan-1-ol
Molecular Weight (g/mol) 340.64
PubChem CID 18431
ChEBI CHEBI:78411
Formula Weight 340.63
Percent Purity 99+%
Physical Form Fine Crystalline Powder or Flakes
Chemical Name or Material Tricosanol, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.