Learn More
Cytiva Tricorn™ Filter Kits
Filter kits for Tricorn™ 5 and 10 columns
Brand: Cytiva 11-0012-54
Code : GE
Additional Details : Weight : 0.00990kg
Description
Kit includes:
- Top and bottom adapters
- Bottom filters
- EPDM O-rings
Specifications
For 10 columns | |
Polyethylene, Ethylene Propylenediene |