Learn More
Cytiva Tricorn™ Bottom Unit
Product Code. 10079229 Shop All Cytiva Products
Click to view available options
For Use With:
For 10 columns
For 5 columns
Unit Size:
Each
Product Code. 10079229
Brand: Cytiva 18115310
Only null left
Add to basket
Add to basket
Designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures
  • Tricorn columns are designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures. They are suitable for use with high performance media, such as Sepharose High Performance, Superdex Prep Grade, and SOURCE.
  • The columns are available with inner diameters of 5 and 10mm, and in lengths from 20 to 200mm. Column lengths of 300 and 600mm are also available for the 10mm diameter columns. The design and dimensions of the columns make them particularly useful for screening and polishing steps in protein purification.
  • Glass tubes coated with plastic film protects individuals in the laboratory from harm if the column is dropped or operated outside its specification.
TRUSTED_SUSTAINABILITY
Specifications
For Use With For 10 columns
Material Ethylene Propylene, PEEK, Polyethylene