missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ TRICONYL MITT Gloves
Description
- 100 % polyamide knitted glove
- White color, PVC dots on one face
- End of the fingers cut
- 5 cm elastic wrist
- Ambidextrous
- The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
- The glove offers maximum comfort and added sturdiness
- Non pilling, the polyamide fabric is suitable for the agrifood industry as well as for precision work
- The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- End of the fingers cut for a better use and handling of small parts
- The PVC dots ensure greater abrasion resistance and a better grip
Specifications
Specifications
| Certifications/Compliance | EN 420-2003 EEC Basic standards, EN 388-2016 Mechanical hazards |
| Description | 13 gauge |
| Material | Polyamide |
| Size | 9 |
| Length (Metric) | 24 cm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?