missing translation for 'onlineSavingsMsg'
Learn More

Triclosan, TRC

Product Code. 30456639
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456639 10 g 10g
30456273 100 mg 100mg
30455896 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456639 Supplier Toronto Research Chemicals Supplier No. TRCT77425010G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3380-34-5
Molecular Formula C12 H7 Cl3 O2
Molecular Weight (g/mol) 289.54
Synonym Triclosan, Phenol, 5-chloro-2-(2,4-dichlorophenoxy)-, 5-Chloro-2-(2,4-dichlorophenoxy)phenol, 2,2'-Oxybis(1',5'-dichlorophenyl-5-chlorophenol), 2,4,4'-Trichloro-2'-hydroxydiphenyl ether, 2,4,4'-Trichloro-2'-hydroxyldiphenyl ether, 2-(2,4-Dichlorophenoxy)-5-chlorophenol, 2-Hydroxy-2',4,4'-trichlorodiphenyl ether, 2',4,4'-Trichloro-2-hydroxydiphenyl ether, 2',4',4-Trichloro-2-hydroxydiphenyl ether, 2'-Hydroxy-2,4,4'-trichlorodiphenyl ether, 3-Chloro-6-(2,4-dichlorophenoxy)phenol, 4-Chloro-2-hydroxyphenyl 2,4-dichlorophenyl ether, Anvasan AM 110, Anvasan AM 110 new, Aquasept, Bacti-Stat soap, CH 3565, CH 3635, Cansan TCH, DP 300, Gamophen, Germicide CA, Invasan, Invasan DP 300R, Irgacare MP, Irgacide LP 10, Irgaguard B 1000, Irgaguard B 1325, Irgasan, Irgasan CH 3565, Irgasan DP 30, Irgasan DP 300, Irgasan DP 3000, Irgasan DP 400, Irgasan PE 30, Irgasan PG 60, Microban, Microban Additive B, Microban B, Microfresh 9200-200, Microfresh Liquid Formulation 9200-200, NM 100, Oletron, Sanitized XTX, Sapoderm, SterZac, TCCP, THDP, Tinosan AM 100, Tinosan AM 110, Ultra-Fresh NM 100, Ultra-Fresh NM-V 2, Vinyzene DP 7000, Yujiexin, Zilesan UW
IUPAC Name 5-chloro-2-(2,4-dichlorophenoxy)phenol
SMILES Oc1cc(Cl)ccc1Oc2ccc(Cl)cc2Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H7Cl3O2/c13-7-1-3-11(9(15)5-7)17-12-4-2-8(14)6-10(12)16/h1-6,16H
Formula Weight 287.9512
Percent Purity >95%
Chemical Name or Material Triclosan
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.