missing translation for 'onlineSavingsMsg'
Learn More

Triclosan O-beta-D-Glucuronide Sodium Salt, TRC

Product Code. 30453133
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
10 mg
missing translation for 'unitSize':
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30453133 10 mg 10mg
30456285 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30453133 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCT77426010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triclosan O-beta-D-Glucuronide Sodium Salt
Analyte or Component Names Triclosan O-beta-D-Glucuronide Sodium Salt
Purity Grade Notes HPLC
Shipping Condition Room Temperature
Molecular Formula C18H14Cl3NaO8
Synonym 5-Chloro-2-(2,4-dichlorophenoxy)phenyl β-D-Glucopyranosiduronic Acid Sodium Salt, Irgasan DP 300 Glucuronide Sodium Salt
InChI Formula InChI=1S/C18H15Cl3O8.Na/c19-7-1-3-10(9(21)5-7)27-11-4-2-8(20)6-12(11)28-18-15(24)13(22)14(23)16(29-18)17(25)26;/h1-6,13-16,18,22-24H,(H,25,26);/q;+1/p-1/t13-,14-,15+,16-,18+;/m0./s1
Solution Type Neat
SMILES ClC1=C(OC2=CC=C(Cl)C=C2O[C@@H]3O[C@H](C(O[Na])=O)[C@@H](O)[C@H](O)[C@H]3O)C=CC(Cl)=C1
Industry Type Forensics
Molecular Weight (g/mol) 487.65
Quantity 10 mg
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.