missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triclabendazole Sulfoxide-D3, TRC
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Specifications
Specifications
| Chemical Name or Material | Triclabendazole Sulfoxide-D3 |
| Molecular Formula | C14H6D3Cl3N2O2S |
| Quantity | 1 mg |
| InChI Formula | InChI=1S/C14H9Cl3N2O2S/c1-22(20)14-18-9-5-8(16)12(6-10(9)19-14)21-11-4-2-3-7(15)13(11)17/h2-6H,1H3,(H,18,19)/i1D3 |
| SMILES | ClC1=CC=CC(OC2=C(Cl)C=C(N=C(S(C([2H])([2H])[2H])=O)N3)C3=C2)=C1Cl |
| IUPAC Name | 5-chloro-6-(2,3-dichlorophenoxy)-2-((methyl-d3)sulfinyl)-1H-benzo[d]imidazole |
| Molecular Weight (g/mol) | 378.68 |
| Formula Weight | 376.96 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction