missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triclabendazole Sulfone-D3, TRC
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Specifications
Specifications
| Chemical Name or Material | Triclabendazole Sulfone D3 (S-methyl-D3) |
| Molecular Formula | C14 D3 H6 Cl3 N2 O3 S |
| Quantity | 10 mg |
| Synonym | Triclabendazole Sulfone D3 (S-methyl-D3), Triclabendazole sulfone-d3, 6-Chloro-5-(2,3-dichlorophenoxy)-2-[(methyl-d3)sulfonyl]-1H-benzimidazole, 6-Chloro-5-(2,3-dichlorophenoxy)-2-(trideuteriomethylsulfonyl)-1H-benzimidazole, Triclabendazole Sulfone-D3 |
| InChI Formula | InChI=1S/C14H9Cl3N2O3S/c1-23(20,21)14-18-9-5-8(16)12(6-10(9)19-14)22-11-4-2-3-7(15)13(11)17/h2-6H,1H3,(H,18,19)/i1D3 |
| SMILES | [2H]C([2H])([2H])S(=O)(=O)c1nc2cc(Cl)c(Oc3cccc(Cl)c3Cl)cc2[nH]1 |
| IUPAC Name | 5-chloro-6-(2,3-dichlorophenoxy)-2-(trideuteriomethylsulfonyl)-1H-benzimidazole |
| Molecular Weight (g/mol) | 394.68 |
| Formula Weight | 392.9588 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction