missing translation for 'onlineSavingsMsg'
Learn More

Triclabendazole, MedChemExpress

Product Code. 16452186
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452186 500 mg 500mg
16448396 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16452186 Supplier Medchem Express Supplier No. HYB0621
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Triclabendazole(CGA89317) is a benzimidazole, it binds to tubulin impairing intracellular transport mechanisms and interferes with protein synthesis.

Triclabendazole(CGA89317) is a benzimidazole, it binds to tubulin impairing intracellular transport mechanisms and interferes with protein synthesis.

Specifications

Chemical Name or Material Triclabendazole
CAS 68786-66-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H9Cl3N2OS
Quantity 500 mg
Synonym CGA89317
Solubility Information DMSO : 100 mg/mL (278.04 mM; Need ultrasonic)
SMILES CSC1=NC2=CC(OC3=CC=CC(Cl)=C3Cl)=C(Cl)C=C2N1
Molecular Weight (g/mol) 359.66
Formula Weight 359.66
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.