missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tricine (White Crystals), Fisher BioReagents
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Poly Bottle; 100g
This biological buffer has a usable pH range of 7.4 to 8.8.
Specifications
Specifications
| Absorbance | At 260nm (0.1M solution) is ≤0.04At 280nm (0.1M solution) is ≤0.02 |
| Chemical Name or Material | Tricine |
| CAS | 4-1-5704 |
| Melting Point | 182°C |
| Color | White |
| pH | 4.6 to 6.0 |
| Physical Form | Powder or Solid |
| Quantity | 100 g |
| Assay Percent Range | ≥98 % |
| Identification | Pass Test |
| Show More |
Safety and Handling
Product Identifier
- Tricine
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction