missing translation for 'onlineSavingsMsg'
Learn More

Tricine Buffer, MP Biomedicals

Product Code. 11443511 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g

Product Code. 11443511

Brand: MP Biomedicals 0210311225

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 4-1-5704
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
Synonym N-[2-Hydroxy-1, 1-bis(hydroxymethyl) ethyl] glycine, N-Tris-[hydroxymethyl]methylglycine
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Color White
Physical Form Powder
Quantity 25 g
Formula Weight 179.2
Chemical Name or Material Tricine Buffer, Cell Biology and Analysis, Molecular Biology
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.