missing translation for 'onlineSavingsMsg'
Learn More

Tricine Biological buffer MP Biomedicals

Product Code. 11473511 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kg
Unit Size:
1kg

Product Code. 11473511

Brand: MP Biomedicals 0210311291

Please to purchase this item. Need a web account? Register with us today!

Specifications

CAS 4-1-5704
Molecular Formula C6H13NO5
Quantity 1 kg
Synonym tricine, n-tris hydroxymethyl methylglycine, n-tri hydroxymethyl methyl glycine, n-tris hydroxymethyl methyl glycine, glycine, n-2-hydroxy-1,1-bis hydroxymethyl ethyl, n-2-hydroxy-1,1-bis hydroxymethyl ethyl glycine, unii-w12lh4v8v3, n-tris-hydroxymethyl methylglycine
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
SMILES C(C(=O)O)NC(CO)(CO)CO
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
Molecular Weight (g/mol) 179.17
PubChem CID 79784
ChEBI CHEBI:39063
Formula Weight 179.2
Chemical Name or Material Tricine, Biological buffer
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.