missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tricine, 98%, for biochemistry

Product Code. 10348603 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
250g
25g
This item is not returnable. View return policy

Product Code. 10348603

Brand: Thermo Scientific Chemicals 172640010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4-1-5704
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
Synonym N-(2-Hydroxy-1, 1-bis(hydroxymethyl)ethyl)glycine, N-[Tris(hydroxymethyl)methyl]glycine
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Absorbance (5% aq. soln.) (1 cm cell)
Absorbance Conditions (5% aq. soln.) (1 cm cell)
Melting Point 180.0°C to 184.0°C
Color White
pH 4.8 to 5.4 (0.5M aq. soln. at 20°C)
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 98%
Infrared Spectrum Authentic
Loss on Drying 1% max. (120°C)
Linear Formula (HOCH2)3CNHCH2CO2H
Packaging Plastic bottle
Merck Index 15, 9816
Solubility Information Solubility in water: soluble.
Formula Weight 179.17
Percent Purity 98%
Grade Biochemical
Chemical Name or Material Tricine, 98%
Show More Show Less
Product Identifier
  • Tricine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.