missing translation for 'onlineSavingsMsg'
Learn More

Tricine, 98+%, Thermo Scientific Chemicals

£19.50 - £164.00

Chemical Identifiers

CAS 5704-04-1
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
Synonym N-(Trishydroxymethylmethyl)glycine
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11497696
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14695.14
25 g
£19.50
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11407706
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14695.22
100 g
£40.80
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11417706
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14695.36
500 g
£164.00
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Good's buffers Tricine is used as an electrophoresis buffer and involved in the separation of low molecular weight proteins and peptides. It operates in the pH range between 7.4 and 8.8. It acts as an effective scavenger of hydroxyl radicals during the study of radiation-induced membrane damage. It is also used in resuspension of cell pellets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Good′s buffers Tricine is used as an electrophoresis buffer and involved in the separation of low molecular weight proteins and peptides. It operates in the pH range between 7.4 and 8.8. It acts as an effective scavenger of hydroxyl radicals during the study of radiation-induced membrane damage. It is also used in resuspension of cell pellets.

Solubility
Soluble in water.

Notes
pH range: 7.8-8.8Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

5704-04-1
179.17
SEQKRHFRPICQDD-UHFFFAOYSA-N
79784
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
C6H13NO5
MFCD00004277
N-(Trishydroxymethylmethyl)glycine
CHEBI:39063
C(C(=O)O)NC(CO)(CO)CO

Specifications

5704-04-1
C6H13NO5
1937804
N-(Trishydroxymethylmethyl)glycine
SEQKRHFRPICQDD-UHFFFAOYSA-N
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
79784
179.17
Tricine
∼183°C (decomposition)
MFCD00004277
14,9651
Soluble in water.
C(C(=O)O)NC(CO)(CO)CO
179.17
CHEBI:39063
≥98%
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 227-193-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only