missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Tricine
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- pKa 25°C: 8.1-8.2
- Melting Range: 182-188°C
Specifications
Specifications
| Chemical Name or Material | Tricine, Ultra Pure |
| CAS | 4-1-5704 |
| Quantity | 500 g |
| Molecular Formula | C6H13NO5 |
| Grade | Ultra Pure |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction