missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Tricine
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
Description
- Tricine is used as buffer component for separation of low molecular weight peptides.
- Tricine can be used in cryopreservation medium for the preservation of tissues and organs.
- Cryopreservation depends on the physical and chemical characteristics of the preservation medium used.
- The pH values and pK values for tricine/DMSO mixtures has been reported down to -20°C.
- Tricine has been found to be an efficient scavenger of hydroxyl radicals in a study of radiation-induced membrane damage.
- Tricine is typically the buffer of choice in SDS-PAGE systems when separating proteins in the range of 1 to 100 kDa.
Specifications
Specifications
| Color | Clear, colorless |
| Description | N-Tris-[hydroxymethyl]methylglycine, N-[2-Hydroxy-1,1-bis(hydroxymethyl) ethyl] glycine |
| Physical Form | Powder |
| Quantity | 25 g |
| Solubility Information | Very soluble in water (25% w/v - clear, colorless solution). |
| Percent Purity | ≥99% |
| Grade | Electrophoresis |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction