missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Trichostatin A

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Trichostatin A is a selective and potent inhibitor of class I and II histone deacetylases (HDAC) (Ki = 3.4 nM, IC50 values are 4.99 nM, 5.21 nM, 16.4 nM, 24.3 nM, 27.6 nM for HDAC1, 3, 6, 10 and 4, respectively). Trichostatin A induces accelerated dedifferentiation of primordial germ cells (PGCs) into embryonic germ (EG) cells. Trichostatin A potently inhibits a range of breast cancer and melanoma cell lines (IC50 = 29-400 nM) and induces apoptosis in prostate cancer cells. In an animal model of Huntington's disease, inhibition of HDAC by Trichostatin A rescues memory deficits and increases expression of CREB/CBP target genes in mutant mice. Activates autophagy, increases vesicular transport of BDNF (Cat. No. 2837) and is active in vivo.
Specifications
Specifications
| CAS | 58880-19-6 |
| Quantity | 1 mg |
| Molecular Formula | C17H22N2O3 |
| Formula Weight | 302.37 |
| Purity | 97% |
| Molecular Weight (g/mol) | 302.37 |
| Chemical Name or Material | Trichostatin A |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction