missing translation for 'onlineSavingsMsg'
Learn More

Trichostatin A, MedChemExpress

Product Code. 16490226
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490226 2 mg 2mg
16478907 5 mg 5mg
16472437 50 mg 50mg
16416328 10 mg 10mg
16413466 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16490226 Supplier Medchem Express Supplier No. HY15144
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Trichostatin A (TSA) is a potent and specific inhibitor of HDAC class I/II, with an IC50 value of 1.8 nM for HDAC.

Trichostatin A (TSA) is a potent and specific inhibitor of HDAC class I/II, with an IC50 value of 1.8 nM for HDAC.

Specifications

Chemical Name or Material Trichostatin A
Physical Form Solid
CAS 58880-19-6
Color Light Nude
Quantity 2 mg
Purity Grade Notes Research
Molecular Formula C17H22N2O3
For Use With (Application) Cancer-programmed cell death
Synonym TSA
Solubility Information DMSO : 25 mg/mL (82.68 mM; Need ultrasonic)
SMILES CN(C1=CC=C(C=C1)C([C@@H](/C=C(/C=C/C(NO)=O)C)C)=O)C
Molecular Weight (g/mol) 302.37
Formula Weight 302.37
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.