missing translation for 'onlineSavingsMsg'
Learn More

Trichostatin A, MedChemExpress

Product Code. 16413466
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16413466

Brand: Medchem Express HY15144

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Trichostatin A (TSA) is a potent and specific inhibitor of HDAC class I/II, with an IC50 value of 1.8 nM for HDAC.

Trichostatin A (TSA) is a potent and specific inhibitor of HDAC class I/II, with an IC50 value of 1.8 nM for HDAC.

Specifications

Chemical Name or Material Trichostatin A
Physical Form Solid
CAS 58880-19-6
Color Light Nude
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C17H22N2O3
For Use With (Application) Cancer-programmed cell death
Synonym TSA
Solubility Information DMSO : 25 mg/mL (82.68 mM; Need ultrasonic)
SMILES CN(C1=CC=C(C=C1)C([C@@H](/C=C(/C=C/C(NO)=O)C)C)=O)C
Molecular Weight (g/mol) 302.37
Formula Weight 302.37
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.